Learn More
Invitrogen™ Vybrant™ DiI Cell-Labeling Solution
Lipophilic membrane stain that diffuses laterally to stain the entire cell
Brand: Invitrogen™ V22885
182.40 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : 50
Additional Details : Weight : 0.00100kg Transport : UN number : 1170 Chem class : 3 Pack group : II
Description
DiI is weakly fluorescent until incorporated into membranes. This orange-red-fluorescent dye, which is spectrally similar to tetramethylrhodamine, is often used as a long-term tracer for neuronal and other cells. DiI is also available as a solid (D-282), as a paste (N-22880) or as crystals (D-3911).
Cell Analysis, Cell Proliferation, Cell Tracing & Tracking, Cell Viability, Proliferation & Function, General Cell Tracing, Microbial Tracking, Neuronal Tracing
Order Info
Shipping Condition: Room Temperature
Specifications
Red-Orange | |
Fluorescence Microscope, Flow Cytometer | |
Vybrant™ | |
Fluorescent Dye | |
Cell Membranes, Lipids |
Store at room temperature and protect from light. | |
Fluorescent | |
Room Temperature | |
Cell Labeling |
Safety and Handling
- Vybrant DiI Cell-Labeling Solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P243-Take precautionary measures against static discharge.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : ethanol
For Research Use Only. Not for use in diagnostic procedures.