Learn More
Invitrogen™ XTT (2,3-Bis-(2-Methoxy-4-Nitro-5-Sulfophenyl)-2H-Tetrazolium-5-Carboxanilide)
Redox-based viability probe
Brand: Invitrogen™ X6493
Code : 50
Additional Details : Weight : 0.00450kg
Description
XTT is used to assess cell viability as a function of redox potential. Actively respiring cells convert the water-soluble XTT to a water-soluble, orange colored formazan product. Unlike MTT, XTT does not require solubilization prior to quantitation, thereby reducing the assay time in many viability assay protocols.
- Compatible Cells: Eukaryotic Cells
- Platform: Microplate Reader
- Detection Method: Fluorescent
- Format: 96-well plate
- Sensitivity of XTT reduction assay is reported to be similar to or better than that of the MTT reduction assay
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Nitro-Oxidative Stress, Reactive Oxygen Species
Order Info
Shipping Condition: Room temperature
Specifications
Eukaryotic Cells | |
Colorimetric | |
XTT | |
96-well plate |
Store in freezer -5°C to -30°C. | |
Microplate Reader | |
Other Label(s) or Dye(s) | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.