Learn More
Invitrogen™ Zymosan A (S. cerevisiae) BioParticles™, Alexa Fluor™ 488 conjugate
Study phagocytosis by fluorescence microscopy, quantitative spectrofluorometry, and flow cytometry
Brand: Invitrogen™ Z23373
176.67 GBP valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
- Fluorescence of fluorescein-labeled BioParticles conjugates, partially quenched in acidic environments, fluorescence of Alexa Fluor, BODIPY™ FL, tetramethylrhodamine and Texas Red™ dye conjugates is uniformly intense over the pH range from 3 to 10
- Label (Ex/Em): Alexa Fluor488 (approx. 495/519nm)
- Particle: Zymosan (S. cerevisiae)
- Provided as lyophilized powders
- Approximately 3 x 108 E. coli or S. aureus particles/mg solid and approximately 2 x 107 zymosan particles/mg solid
- Conjugates can be reconstituted in buffer of choice for use in phagocytosis assays
- Fluorescence of BioParticles conjugates that are bound to surface of cell (but not internalized) can be quenched by ethidium bromide, trypan blue, or other quenchers
- Addition to cellular applications, fluorescent BioParticles conjugates may be effective as flow cytometry calibration references when sorting bacteria and yeast mutants
- Small particles may be useful references for light scattering studies because their sizes and shapes differ in characteristic ways
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room Temperature
Specifications
Store in freezer (-5 to -30°C) and protect from light. | |
Fluorescence Microscope | |
Alexa Fluor™ 488 | |
3 to 10 | |
2 mg | |
S. cerevisiae |
Fluorescence | |
Supplement | |
Lyophilized Powder | |
Alexa Fluor™, BioParticles™ | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.