Learn More
Invitrogen™ Zymosan A S. cerevisiae BioParticles™, unlabeled
Nonfluorescent products are useful in phagocytosis assays, both as controls and for custom labeling with reactive dyes
Brand: Invitrogen™ Z2849
88.90 GBP valid until 2025-06-30
Use promo code "24074" to get your promotional price.
Description
- Label (Ex/Em): Unlabeled
- Particle: Zymosan (S. cerevisiae)
- Provided as lyophilized powders
- Approximately 3 x 108 S. aureus particles per mg solid and approximately 2 x 107 zymosan particles per mg solid
- Conjugates can be reconstituted in the buffer of choice for use in phagocytosis assays
Cell Analysis, Cell Viability, Proliferation and Function, Cellular Imaging, Endocytosis, Exocytosis and Phagocytosis, Flow Cytometry, Flow Cytometry Endocytosis, Exocytosis and Phagocytosis Assays, Flow Cytometry of Cellular Processes, Immunofluorescence (IF), Immunofluorescence Staining and Detection, Ionic Homeostasis and Signaling, pH Detection
Order Info
Shipping Condition: Room Temperature

Specifications
Store in freezer (-5 to -30°C). | |
Fluorescence Microscope | |
Unconjugated | |
BioParticles™ | |
Room Temperature |
Fluorescence | |
Bioparticle Conjugate | |
Solid | |
100 mg | |
S. cerevisiae |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.