Learn More
Molecular Shapes Model Kit
Product Code. 12889515
Encompass_Preferred
Click to view available options
:
Includes: Linear Hydrogen Chloride, Linear Barylium Chloride, Bent Water, Trigonal Planar Boron Trihydride, Pyramidal Ammonia, Tetrahedral Methane, Trigonal Bipyramidal Phosphorus Pentachloride, Octahedral Sulphur Hexafloride
Product Code. 12889515
Only null left
Add to basket
Add to basket
Specifications
For Use With (Application) Demonstrate atomic structure and function
Includes 13 Hydrogen (white), 7 Chlorine (green), 9 Fluorine (light green), 1 Metal (Beryllium) (gray), 1 Boron (beige), 1 Chlorine (light green), 1 Oxygen (red), 1 Nitrogen (blue), 1 Carbon (black), 1 Phosphorus (purple), 1 Sulphur (yellow), 26 Sigma bonds (grey), 6 Lone pair orbital (beige), 6 Protonated lone pair orbital (beige), 6 Short Link (white)
Product Type Molecular Shapes - Electron Repulsion Theory Molyorbital 8 Model Collection Set