Learn More
Molecular Sieve Dehydrate with Indicator, Honeywell Fluka™
Rod Size 1/16 In., with Indicator for Drying Solvents
Brand: Honeywell Fluka 69828-1KG
Code : NEW
Additional Details : Weight : 1.00000kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Poly bottle | |
1 kg | |
Molecular Sieve Dehydrate Fluka with indicator for drying solvents | |
UN3077 |