missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 13X, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Specifications
Specifications
Packaging | Drum with inliner |
Quantity | 5 kg |
CAS | 63231-69-6 |
Linear Formula | Na86[AlO2)86(SiO2)106] · xH2O |
MDL Number | MFCD00131613 |
UN Number | NONH for all modes of transport |
IUPAC Name | Molecular sieves 13X |
Chemical Name or Material | Molecular Sieves, 13X |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molecular Sieves, 13X, Honeywell™ Fluka™ >
Spot an opportunity for improvement?Share a Content Correction