missing translation for 'onlineSavingsMsg'
Learn More

Molecular Sieves, 3 Å, 4 to 8mesh, Honeywell Fluka™

Product Code. 15671840
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. Rückgaberichtlinie anzeigen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Beads, 4-8 mesh

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 308080-99-1
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.135
MDL Number MFCD00131613
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3]

Spezifikation

Mesh Size 4 to 8mesh
Quantity 5 kg
Linear Formula KnNa12-n[(AlO2)12(SiO2)12] · xH2O
UN Number NONH for all modes of transport
Chemical Name or Material Molecular Sieves, 3 Å
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'