missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 3 Å, 8 to 12mesh, Honeywell Fluka™
Beads, 8-12 mesh
£105.47 - £329.59
Chemical Identifiers
CAS | 308080-99-1 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.135 |
MDL Number | MFCD00131613 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
IUPAC Name | aluminum;sodium;dioxido(oxo)silane |
SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15681840
|
Honeywell-Fluka
208582-1KG |
1 kg |
£105.47
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15691840
|
Honeywell-Fluka
208582-5KG |
5 kg |
£329.59
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
308080-99-1 | |
202.135 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane |
AlNaO6Si2 | |
MFCD00131613 | |
19758701 | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Specifications
8 to 12mesh | |
AlNaO6Si2 | |
MFCD00131613 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
aluminum;sodium;dioxido(oxo)silane | |
19758701 |
308080-99-1 | |
KnNa12-n[(AlO2)12(SiO2)12] · xH2O | |
NONH for all modes of transport | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] | |
202.135 | |
Molecular Sieves, 3 Å |