missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 3A, Powder, Honeywell Fluka™
Powder
£37.51 - £108.91
Chemical Identifiers
CAS | 308080-99-1 |
---|---|
Molecular Formula | AlNaO6Si2 |
Molecular Weight (g/mol) | 202.135 |
MDL Number | MFCD00131613 |
InChI Key | URGAHOPLAPQHLN-UHFFFAOYSA-N |
PubChem CID | 19758701 |
SMILES | [O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
308080-99-1 | |
202.135 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] |
AlNaO6Si2 | |
MFCD00131613 | |
19758701 |
Specifications
White | |
3Å | |
powder | |
AlNaO6Si2 | |
MFCD00131613 | |
URGAHOPLAPQHLN-UHFFFAOYSA-N | |
Molecular sieves 3A | |
19758701 |
>2g/cm3 | |
308080-99-1 | |
>400°C | |
KnNa12-n[(AlO2)12(SiO2)12] · xH2O | |
NONH for all modes of transport | |
[O-][Si](=O)[O-].[O-][Si](=O)[O-].[Na+].[Al+3] | |
202.135 | |
Molecular sieves, 3Å |