missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 4 Å, 4 to 8mesh, Honeywell Fluka™
Click to view available options
Quantity:
500 g
1 kg
5 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Mesh Size | 4 to 8mesh |
Quantity | 1 kg |
Linear Formula | Na12[(AlO2)12(SiO2)12] · xH2O |
UN Number | NONH for all modes of transport |
Chemical Name or Material | Molecular Sieves, 4 Å |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'