missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular sieves 4A, 10 to 18 mesh, Thermo Scientific Chemicals
£51.70 - £224.00
Chemical Identifiers
CAS | 70955-01-0 |
---|---|
Molecular Formula | C20H25FN2O8 |
Molecular Weight (g/mol) | 440.42 |
MDL Number | MFCD00131613 |
InChI Key | FJUZEZRZKVMAMD-UHFFFAOYNA-N |
PubChem CID | 73906282 |
IUPAC Name | Molecular sieves |
SMILES | COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10542261
|
Thermo Scientific Acros
264755000 |
500 g | Glass bottle |
£51.70
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10013350
|
Thermo Scientific Acros
264750050 |
5 kg | Plastic drum |
£224.00
5kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Drying agentChemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
Molecular sieves |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Specifications
Beige | |
< 1.5% (550°C, 2 hrs.) | |
70955-01-0 | |
C20H25FN2O8 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
Molecular sieves | |
73906282 |
680 to 760g/L | |
> 20% (50% rH, 20°C, 24 h) | |
Granules | |
MFCD00131613 | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 | |
Molecular sieves 4A |
Safety and Handling
HYGROSCOPIC
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title