missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1000 g
5000 g
Description
Molecular sieves, 4A, 3-5mm (0.12-0.20in) beads are useful in general gas drying applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Sensitivity | Hygroscopic |
Quantity | 250 g |
Solubility Information | Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone. |
Chemical Name or Material | Molecular sieves, 4A, 3 to 5mm (0.12 to 0.20 in) beads |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'