missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, 8 to 12 mesh

Product Code. p-44873 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
5 kg
25 kg
Packaging:
Fibre Drum
Glass bottle
Plastic drum
Unit Size:
25kg
500g
5kg
This item is not returnable. View return policy

Product Code. 10216450

Brand: Thermo Scientific Acros 197275000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 70955-01-0 | C20H25FN2O8 | 440.42 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color Beige
Bulk Density 680 to 760g/L
Packaging Glass bottle
Residual Water 1.5% max.
Water Adsorption 20% min.
Quantity 500 g
Physical Form Beads
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
Chemical Name or Material Molecular sieves 4A, 8 to 12 mesh

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.