missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. pellets, Thermo Scientific Chemicals

Product Code. 11357758
Click to view available options
Quantity:
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Molecular sieves, 5A, 3-4mm (0.12-0.16in) dia. Pellets is used as an sorbent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Color Brown
Sensitivity Hygroscopic
Quantity 1 kg
Physical Form Pellet
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves 5A, 3 to 4mm (0.12 to 0.16 in.) dia. pellets

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'