missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 5A, 8 to 12 mesh, Thermo Scientific Chemicals

Product Code. 10451241
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Glass bottle
Plastic drum

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Color Beige to Gray or Brown
Bulk Density 680 to 760g/L
Packaging Plastic drum
Residual Water 1.5% max. (550°C, 2 hrs.)
Water Adsorption 20% min. (50% rH, 20°C, 24 Hrs.)
Quantity 5 kg
Physical Form Granules
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Chemical Name or Material Molecular sieves 5A, 5A, 8 to 12 mesh

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'