missing translation for 'onlineSavingsMsg'
Learn More
Molybdenite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific Chemicals Product Code.: 11335357

Molybdenite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific Chemicals

Product Code. 11335357
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11335357

Brand: Thermo Scientific Alfa Aesar 042642.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals.

Molybdenite, naturally occurring mineral, grains, is used for grain mounts, XRD and microprobe standards, as well as identification of unknown minerals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1309-56-4
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
MDL Number MFCD00003470
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z
PubChem CID 14823
ChEBI CHEBI:30704
IUPAC Name bis(sulfanylidene)molybdenum
SMILES S=[Mo]=S

Specifications

Quantity 25 g
Solubility Information Soluble in sulfuric and nitric acid
Formula Weight 160.07
Packaging Glass bottle
Chemical Name or Material Molybdenite

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenite, naturally occurring mineral, grains, ≈0.06 to 0.19 in., Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.