missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals

Product Code. 11306588
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molybdenum carbide is used in the production of solid molybdenum carbide grades of hard metal in the manufacture of mixed crystal. It used as coating material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12069-89-5
Molecular Formula CMo2
Molecular Weight (g/mol) 203.91
MDL Number MFCD00014218
InChI Key QIJNJJZPYXGIQM-UHFFFAOYSA-N
Synonym dimolybdenum carbide, methane dimolybdenum, molybdenum carbide mo2c, molybdenum carbide trace metals basis 100g
IUPAC Name (molybdeniomethylidene)molybdenum
SMILES [Mo]=C=[Mo]

Specifications

Color White
Melting Point 2687°C
Appearance White powder
Assay Percent Range 99.5% (metals basis)
Quantity 500 g
Solubility Information Insoluble in water.
Formula Weight 203.91
Odor Odorless
Form Powder
Chemical Name or Material Molybdenum carbide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'