missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 15477345

Molybdenum foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15477345
300 x 600 mm
Click to view available options
Quantity:
100 x 100 mm
150 x 300 mm
300 x 600 mm
This item is not returnable. View return policy

Product Code. 15477345

Brand: Thermo Scientific Alfa Aesar 046943.HX

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 300 x 600 mm
Percent Purity 99.95%
Weight ≈5.1g/100x100mm
Form Foil
Assay (metals basis)
Chemical Name or Material Molybdenum foil, 0.05mm (0.002 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberQ20I041 Certificate TypeCertificate of Analysis Date17/11/2022 Catalog Number046943.GH,046943.HX,046943.HW
Lot NumberP15I019 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number046943.GH,046943.HX,046943.HW
Lot NumberQ18D002 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number046943.GH,046943.HX,046943.HW
Lot NumberJ04X064 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number046943.GH,046943.HX,046943.HW
Lot NumberS30B017 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number046943.GH,046943.HX,046943.HW

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum foil, 0.05mm (0.002in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.