missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum foil, 0.1mm (0.004in) thick, 99.95% (metals basis), Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
100 x 150 mm
150 x 300 mm
300 x 600 mm
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Spécification
Color | Gray |
Appearance | Dark gray foil |
Density | 10.22g/cm3 at 20°C |
Assay Percent Range | 99.95% (Metals basis) |
Quantity | 100 x 150 mm |
Formula Weight | 95394 |
Odor | Odorless |
Form | Foil |
Chemical Name or Material | Molybdenum |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'