missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire, Thermo Scientific Chemicals Product Code.: 15477265

Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire, Thermo Scientific Chemicals

Product Code. 15477265
75 x 75 mm
Click to view available options
Quantity:
75 x 75 mm
150 x 150 mm
300 x 300 mm
This item is not returnable. View return policy

Product Code. 15477265

Brand: Thermo Scientific Alfa Aesar 046606.FL

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 75 x 75 mm
Mesh Size 40 Mesh
Form Wire Cloth
Chemical Name or Material Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU19K022 Certificate TypeCertificate of Analysis Date27/09/2024 Catalog Number046606.GT,046606.FL
Lot NumberQ03K022 Certificate TypeCertificate of Analysis Date19/04/2024 Catalog Number046606.GT,046606.FL
Lot NumberU09J048 Certificate TypeCertificate of Analysis Date16/08/2023 Catalog Number046606.GT,046606.HB,046606.FL
Lot NumberR10I021 Certificate TypeCertificate of Analysis Date04/07/2022 Catalog Number046610.FL,046606.GT,046606.HB,046606.FL
Lot NumberY01H024 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number046610.FL,046606.GT,046606.HB,046606.FL

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum gauze, 40 mesh woven from 0.13mm (0.005 in.) dia. wire, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.