missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum hexacarbonyl, 98%, Thermo Scientific Chemicals Product Code.: 10496463

Molybdenum hexacarbonyl, 98%, Thermo Scientific Chemicals

Product Code. 10496463
10 g, Glass bottle
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10496463

Brand: Thermo Scientific Acros 190390100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molybdenum hexacarbonyl, 98%, C6O6Mo, CAS Number-13939-06-5, hexacarbonylmolybdenu, carbon monooxide-molybdenum 6:1, hexacarbonylmolybdenum, may contain up to cr 5g, hexakis carbon monoxide molybdenum, molybdenumhexacarbonyl, hexakis methanidylidyneoxidanium molybdenum, 50g, 100.0, 97.5, CHEBI:30508

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13939-06-5
Molecular Formula C6O6Mo
Molecular Weight (g/mol) 264
MDL Number MFCD00003466
InChI Key KMKBZNSIJQWHJA-UHFFFAOYSA-N
Synonym molybdenumhexacarbonyl, carbon monooxide-molybdenum 6:1, hexakis carbon monoxide molybdenum, hexacarbonylmolybdenu, hexakis methanidylidyneoxidanium molybdenum, hexacarbonylmolybdenum, may contain up to cr 5gShow MoreShow Less
PubChem CID 2724068
ChEBI CHEBI:30508
IUPAC Name carbon monoxide;molybdenum
SMILES [C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Mo]

Specifications

Melting Point 150.0°C
Color White through Yellow
CAS Min % 97.5
CAS Max % 100.0
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 98%
Linear Formula Mo(CO)6
Fieser 02,287; 03,206; 04,346; 09,317; 10,274; 13,194; 01,140; 02,448; 03,325; 04,559; 05,736; 06,652; 07,68; 08,109; 09,113; 15,9
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in benzene, slightly soluble in ether and most oter common, organic solvents
Formula Weight 264
Percent Purity 98%
Infrared Spectrum Authentic
Packaging Glass bottle
Specific Gravity 1.96
Density 1.9600g/mL
Chemical Name or Material Molybdenum hexacarbonyl, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466706 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number190390100,190392500,190390500
Lot NumberA0456759 Certificate TypeCertificate of Analysis Date12/10/2023 Catalog Number190390100,190392500,190390500
Lot NumberA0398793 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number190390100,190392500,190390500
Lot NumberA0403163 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number190390100,190392500,190390500
Lot NumberA0412492 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number190390100,190392500,190390500
compliance-icons
Product Identifier
  • Molybdenum hexacarbonyl
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum hexacarbonyl, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.