missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(II) acetate dimer, Thermo Scientific Chemicals

Product Code. 11353437
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molybdenum(II) acetate dimer is generally used as an intermediate compound in a process to form other quadruply bonded molybdenum compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14221-06-8
Molecular Formula C8H12Mo2O8
Molecular Weight (g/mol) 428.076
MDL Number MFCD00013395
InChI Key DOOLFANBWPPEGQ-UHFFFAOYSA-J
Synonym molybdenum ii acetate, tetrakis mu-acetato-o:o' dimolybdenum, molybdenum 2+ tetraacetate, dimolybdenum tetraacetate, tetra aceto dimolybdenum, molybdenum ii acetate dimer,, molybdenum ii acetate dimer 1g, molybdenum ii acetate dimer
PubChem CID 84269
IUPAC Name molybdenum(2+);tetraacetate
SMILES CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].CC(=O)[O-].[Mo+2].[Mo+2]

Specifications

Quantity 5 g
Sensitivity Air Sensitive
Solubility Information Slightly soluble in organic solvents. Insoluble in water.
Formula Weight 428.06
Physical Form Crystalline
Chemical Name or Material Molybdenum(II) acetate dimer

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.