missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Product Code. 11408640
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18868-43-4
Molecular Formula MoO2
Molecular Weight (g/mol) 127.948
MDL Number MFCD00014223
InChI Key QXYJCZRRLLQGCR-UHFFFAOYSA-N
Synonym molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
PubChem CID 29320
IUPAC Name dioxomolybdenum
SMILES O=[Mo]=O

Specifications

Color Brown to Black
Physical Form Powder
Quantity 25 g
Assay Percent Range 99% (metals basis)
Formula Weight 127.94
Percent Purity 99%
Odor Odorless
Density 6.47 g/mL
Assay (metals basis)
Chemical Name or Material Molybdenum (IV) oxide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.