missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) selenide, 99.9% (metals basis)

Product Code. p-4408165
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11302208

Brand: Thermo Scientific Alfa Aesar 013112.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12058-18-3 | MoSe2 | 253.892 g/mol

Molybdenum(IV) selenide is used in transistors and photodetectors. It is also used as a solid lubricant. Further, it is used in piezoelectric and solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12058-18-3
Molecular Formula MoSe2
Molecular Weight (g/mol) 253.892
MDL Number MFCD00014216
InChI Key MHWZQNGIEIYAQJ-UHFFFAOYSA-N
Synonym molybdenum diselenide, molybdenum selenide, molybdenum iv selenide, diselanylidenemolybdenum, bis selanylidene molybdenum, molybdenum diselenide, crystal, molybdenum iv selenide trace metals basis 5g
PubChem CID 82894
IUPAC Name bis(selanylidene)molybdenum
SMILES [Se]=[Mo]=[Se]

Specifications

Melting Point >1200°C
Color Gray
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (metals basis)
Solubility Information Insoluble in water.
Formula Weight 253.87
Odor Odorless
Appearance Dark gray powder
Density 6g/cm3
Chemical Name or Material Molybdenum(IV) selenide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.