missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum(IV) sulfide, 98.5%, Thermo Scientific Chemicals Product Code.: 10328160

Molybdenum(IV) sulfide, 98.5%, Thermo Scientific Chemicals

Product Code. 10328160
500 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy

Product Code. 10328160

Brand: Thermo Scientific Acros 215785000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1317-33-5
Molecular Formula MoS2
Molecular Weight (g/mol) 160.07
InChI Key CWQXQMHSOZUFJS-UHFFFAOYSA-N
Synonym molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote zShow MoreShow Less
PubChem CID 14823
ChEBI CHEBI:30704
IUPAC Name bis(sulfanylidene)molybdenum
SMILES S=[Mo]=S

Specifications

Melting Point 2375.0°C
Color Black or Gray
Physical Form Powder
Quantity 500 g
Assay Percent Range 98% min. (MoS2)
Linear Formula MoS2
Merck Index 15,6322
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in aqua regia and sulfuric acid
Formula Weight 160.07
Percent Purity 98.5%
Packaging Glass bottle
Chemical Name or Material Molybdenum(IV) sulfide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0464421 Certificate TypeCertificate of Analysis Date10/07/2024 Catalog Number215785000,215780025,215781000
Lot NumberA0459561 Certificate TypeCertificate of Analysis Date24/01/2024 Catalog Number215785000,215780025,215781000
Lot NumberA0398438 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number215785000,215780025,215781000
Lot NumberA0392977 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number215785000,215780025,215781000
Lot NumberA0448654 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number215785000,215780025,215781000
compliance-icons
Product Identifier
  • Molybdenum(IV) sulfide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
Precautionary Statement
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum(IV) sulfide, 98.5%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.