missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum, plasma standard solution, Specpure™, Mo 10,000μg/ml, Thermo Scientific Chemicals Product Code.: 11379007

Molybdenum, plasma standard solution, Specpure™, Mo 10,000μg/ml, Thermo Scientific Chemicals

Product Code. 11379007
50 mL
Click to view available options
Quantity:
50 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11379007

Brand: Thermo Scientific Alfa Aesar 038726.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Physical Form Liquid
Quantity 50 mL
Packaging Plastic bottle
Chemical Name or Material Molybdenum

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1550408 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number038726.AP,038726.AD,038726.AE
Lot Number1408355 Certificate TypeCertificate of Analysis Date12/12/2023 Catalog Number038726.AP,038726.AD,038726.AE
Lot Number1396472 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number038726.AP,038726.AD,038726.AE
Lot Number1387317 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog Number038726.AP,038726.AD,038726.AE
Lot Number1406591 Certificate TypeCertificate of Analysis Date16/10/2023 Catalog Number038726.AP,038726.AD,038726.AE

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum, plasma standard solution, Specpure™, Mo 10,000μg/ml, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.