missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum plasma standard solution, Thermo Scientific Chemicals, Specpure™
£47.54 - £111.00
Chemical Identifiers
CAS | 7439-98-7 |
---|---|
Molecular Formula | Mo |
Molecular Weight (g/mol) | 95.95 |
MDL Number | MFCD00003465 |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
PubChem CID | 23932 |
ChEBI | CHEBI:28685 |
IUPAC Name | molybdenum |
SMILES | [Mo] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11358997
|
Thermo Scientific Alfa Aesar
|
50 mL |
£47.54
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368997
|
Thermo Scientific Alfa Aesar
038719.AE |
100 mL |
£74.37
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430171
|
Thermo Scientific Alfa Aesar
038719.AP |
500 mL | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
CHEBI:28685 | |
[Mo] |
Mo | |
MFCD00003465 | |
23932 | |
molybdenum |
Specifications
7439-98-7 | |
Mo | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
Liquid |
Colorless | |
MFCD00003465 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
Molybdenum, plasma standard solution, Mo 1000μg/mL |
Safety and Handling
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only