missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11305739

Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11305739
50 x 50 mm
Click to view available options
Quantity:
50 x 50 mm
100 x 100 mm
100 x 500 mm
This item is not returnable. View return policy

Product Code. 11305739

Brand: Thermo Scientific Alfa Aesar 010047.FI

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.95%
Weight ≈64g/50x50mm
Form Plate
Assay (metals basis)
Chemical Name or Material Molybdenum plate, 2.5mm (0.098 in.) thick

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP17L022 Certificate TypeCertificate of Analysis Date19/03/2025 Catalog Number010047.GH,010047.FI,010047.HP
Lot NumberM13L001 Certificate TypeCertificate of Analysis Date15/01/2025 Catalog Number010047.GH,010047.FI,010047.HP
Lot NumberY25K014 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number010047.GH,010047.FI,010047.HP
Lot NumberY19F014 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010047.GH,010047.FI,010047.HP
Lot NumberY13F065 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number010047.GH,010047.FI,010047.HP

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum plate, 2.5mm (0.098in) thick, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.