missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11326727
50 g
Click to view available options
Quantity:
50 g
250 g
1 kg
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Density 10.22
Assay Percent Range 99.95% (metals basis)
Quantity 50 g
Formula Weight 95394
Mesh Size -100 Mesh
Form Powder
Chemical Name or Material Molybdenum
compliance-icons
Product Identifier
  • Molybdenum powder
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P240-Ground/bond container and receiving equipment.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P370+P378-In case of fire: Use to extinguish.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.