missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm, Thermo Scientific Chemicals Product Code.: 11302068

Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm, Thermo Scientific Chemicals

Product Code. 11302068
100 g
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy

Product Code. 11302068

Brand: Thermo Scientific Alfa Aesar 012972.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
UN Number UN3089
Quantity 100 g
Percent Purity 99.9975%
Grade Puratronic™
Mesh Size −22 Mesh
Odor Odorless
Form Powder
Assay (metals basis excluding W), W <1000ppm
Chemical Name or Material Molybdenum powder

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberM16E051 Certificate TypeCertificate of Analysis Date05/01/2023 Catalog Number012972.22,012971.22,012971.06
Lot NumberQ08F056 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012972.22,012971.22,012971.06
Lot Number21827 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012972.22,012971.22,012971.06
Lot Number21252 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number012972.22,012971.22,012971.06
Lot NumberE13T025 Certificate TypeCertificate of Analysis Date29/11/2021 Catalog Number012972.22,012971.22,012971.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum powder, -22 mesh, Puratronic™, 99.9975% (metals basis excluding W), W <1000ppm, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.