Learn More
Molybdenum powder, spherical, -170 mesh, 99.5% (metals basis), Thermo Scientific Chemicals
£36.28 - £357.78
Chemical Identifiers
CAS | 7439-98-7 |
---|---|
Molecular Formula | Mo |
Molecular Weight (g/mol) | 95.95 |
MDL Number | MFCD00003465 |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
Synonym | molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 |
PubChem CID | 23932 |
ChEBI | CHEBI:28685 |
IUPAC Name | molybdenum |
SMILES | [Mo] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11334897
|
Thermo Scientific Alfa Aesar
041919.18 |
50 g |
£36.28
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11354897
|
Thermo Scientific Alfa Aesar
041919.30 |
250 g |
£129.65
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344897
|
Thermo Scientific Alfa Aesar
041919.A1 |
1 kg |
£357.78
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2Show More | |
CHEBI:28685 | |
[Mo] |
Specifications
Gray | |
Mo | |
UN3089 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
99.5% | |
Odorless | |
(metals basis) |
7439-98-7 | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
−170 Mesh | |
Spherical Powder | |
Molybdenum powder |
Safety and Handling
GHS H Statement
H228
Flammable solid.
P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
H228-H360-H371
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-107-2
missing translation for 'rtecsNumber' : QA4680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.