missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis), Thermo Scientific™
Click to view available options
Quantity:
1 Ea.
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Specifications
Color | Gray |
CAS | 12136-78-6 |
Melting Point | 1870°C to 2030°C |
Density | 6.31 g/mL |
Molecular Formula | MoSi2 |
MDL Number | MFCD00151535 |
Quantity | 1 Ea. |
Synonym | molybdenum disilicide, molybdenum silicide, ms silicide, molybdenum-silicide, molybdenum silicide, mo5si3, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 50.8mm 2.0in dia x 6.35mm 0.250in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 3.18mm 0.125in thick, molybdenum silicide sputtering target, 76.2mm 3.0in dia x 6.35mm 0.250in thick |
InChI Key | YXTPWUNVHCYOSP-UHFFFAOYSA-N |
SMILES | [Si]=[Mo]=[Si] |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Molybdenum silicide sputtering target, 76.2mm (3.0 in.) dia. x 6.35mm (0.250 in.) thick, 99.5% (metals basis), Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction