missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 11354468

Molybdenum slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 11354468
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 11354468

Brand: Thermo Scientific Alfa Aesar 042359.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx.2.0g/slug

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Color Gray
Appearance Dark gray slug
Density 10.22g/cm3 at 20°C
Assay Percent Range 99.95% (Metals basis)
Quantity 100 g
Odor Odorless
Form Slug
Chemical Name or Material Molybdenum

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY19K011 Certificate TypeCertificate of Analysis Date20/11/2024 Catalog Number042359.22,042359.14
Lot NumberT29K047 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number042359.22,042359.14
Lot NumberW06K007 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number042359.22,042359.14
Lot NumberW06K008 Certificate TypeCertificate of Analysis Date24/09/2024 Catalog Number042359.22,042359.14
Lot NumberT22K038 Certificate TypeCertificate of Analysis Date16/08/2024 Catalog Number042359.22,042359.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum slug, 6.35mm (0.25in) dia x 6.35mm (0.25in) length, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.