missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum Standard Metal Solution 1000ppm, for AAS, Solution HNO3 0.1M, Fisher Chemical™
Click to view available options
Quantity:
100 mL
500 mL
Specifications
Specifications
Packaging | HDPE plastic bottle |
Quantity | 100 mL |
Color | Colorless |
Physical Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction