missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(V) chloride, 95%, Thermo Scientific Chemicals

Product Code. 10144070
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10241-05-1
Molecular Formula Cl5Mo
Molecular Weight (g/mol) 273.20
MDL Number MFCD00003467
InChI Key GICWIDZXWJGTCI-UHFFFAOYSA-I
Synonym molybdenum v chloride, molybdenum pentachloride, molybdenum chloride mocl5, mocl5, pentachloridomolybdenum, molybdenum 5+ chloride, acmc-1bx8l, ksc491a7h
PubChem CID 61497
ChEBI CHEBI:30635
SMILES [Cl-].[Cl-].[Cl-].[Cl-].[Cl-].[Mo+5]

Specifications

Boiling Point 268°C
Melting Point 194°C
Color Black to Blue or Black-Green
CAS Min % 94.0
CAS Max % 100.0
Physical Form Fine Crystals or Crystalline Powder
Quantity 10 g
Assay Percent Range 94% min. (Argentometry)
Linear Formula MoCl5
UN Number 2508
Fieser 06,412; 01,1015; 02,365; 03,250; 04,428; 05,587; 06,519; 08,443; 09,414; 11,471; 12,434; 13,273; 04,99; 16,95
Solubility Information Solubility in water: reacts with water. Other solubilities: soluble in dry ether,dry alcohol and other,organic solvents
IUPAC Name pentachloromolybdenum
Formula Weight 273.2
Percent Purity 95%
Packaging Glass Bottle
Chemical Name or Material Molybdenum(V) chloride, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Molybdenum(V) chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.