missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals Product Code.: 11373528

Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals

Product Code. 11373528
100 mL
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy

Product Code. 11373528

Brand: Thermo Scientific Alfa Aesar 039159.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Molybdenum(V) isopropoxide is used to prepare novel MoVSbOxtype catalysts for selective isobutane oxidation. It is used to prepare molybdenum Oxide Nanofibers by electrospinning.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 209733-38-0
Molecular Formula C15H35MoO5
Molecular Weight (g/mol) 391.39
MDL Number MFCD00210636
InChI Key HLFWWCCBFJUYJL-UHFFFAOYSA-N
Synonym molybdenum v isopropoxide, acmc-20akdz, pentaisopropoxymolybdenum v, molybdenum 5+ pentakis propan-2-olate
PubChem CID 18475294
IUPAC Name molybdenum;propan-2-olate
SMILES CC(C)O[Mo](OC(C)C)(OC(C)C)(OC(C)C)OC(C)C

Specifications

Color Yellow to Brown
Flash Point 12°C (54°F)
Odor Alcohol-like
Linear Formula Mo[OCH(CH3)2]5
Quantity 100 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Miscible with alcohols,ketones and esters.
Formula Weight 391.04
Percent Purity 99.6%
Concentration or Composition (by Analyte or Components) 5% w/v in isopropanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Molybdenum(V) isopropoxide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberK22M21 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039159.AE,039159.AC
Lot NumberB03Q27 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039159.AE,039159.AC
Lot NumberY12A011 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number039159.AE,039159.AC
Lot NumberY11G018 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039159.AE,039159.AC
Lot NumberY07E060 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number039159.AE,039159.AC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum(V) isopropoxide, 99.6% (metals basis), 5% w/v in isopropanol, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.