missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max

Product Code. 11376927
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11376927

Brand: Thermo Scientific Alfa Aesar 010812.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1313-27-5 | MoO3 | 143.95 g/mol

Its chief application is as an oxidation catalyst and as a raw material for the production of molybdenum metal. It is a Precursor to LAMOX fast ion conductors and superconductors. Molybdenum trioxide has also been suggested as a potential anti-microbial agent, e.g., in polymers. In contact with water, it forms H+ ions that can kill bacteria effectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O

Specifications

Boiling Point 1155°C (sublimation)
Melting Point 795°C
Color White to Gray
Physical Form Powder
Quantity 25 g
UN Number UN3288
Merck Index 14,6239
Solubility Information Soluble at 0.490g/1000 cc water @ 28°C. Soluble in concentrated mineral acids.
Formula Weight 143.94
Percent Purity 99.998%
Grade Puratronic
Odor Odorless
Density 4.692 g/mL
Assay (metals basis excluding W), W 300ppm max.
Chemical Name or Material Molybdenum(VI) oxide
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.