missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W), Thermo Scientific Chemicals

£144.72 - £1262.28

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11352018
SDS View Documents
Thermo Scientific Alfa Aesar
012930.14
25 g
£144.72
25g
Please to purchase this item. Need a web account? Register with us today!
11342018
SDS View Documents
Thermo Scientific Alfa Aesar
012930.22
100 g
£430.14
100g
Please to purchase this item. Need a web account? Register with us today!
11362018
SDS View Documents
Thermo Scientific Alfa Aesar
012930.36
500 g
£1262.28
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

It is used as a selective oxidation catalyst, and as a molybdenum oxidase enzyme. It is a source of molybdenum compounds and molybdenum metal which serves as an additive to steel and corrosion-resistant alloys. It is used for lubricants and lubricant additives. It has applications in process regulators, pigments, corrosion inhibitors, electrochemical devices, displays, and smoke suppressants. It finds use in analytical chemistry, ceramic glazes, enamels, energy storage, and in the manufacture of acrylonitrile through catalytic oxidation of ammonia and propene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c

H319-H335-H351

missing translation for 'dotInformation' : Hazard Class: 6.1, Packing Group: III

missing translation for 'einecsNumber' : 215-204-7

missing translation for 'rtecsNumber' : QA4725000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum(VI) oxide, Puratronic™, 99.9995% (metals basis excluding W), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.