missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals Product Code.: 15496575

Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals

Product Code. 15496575
60 m
Click to view available options
Quantity:
60 m
This item is not returnable. View return policy

Product Code. 15496575

Brand: Thermo Scientific Alfa Aesar 043996.8V

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 7439-98-7
Molecular Formula Mo
MDL Number MFCD00003465
Quantity 60 m
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
SMILES [Mo]
IUPAC Name molybdenum
Molecular Weight (g/mol) 95.95
PubChem CID 23932
ChEBI CHEBI:28685
Percent Purity 99.95%
Weight ≈1.1g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Molybdenum wire, 0.375mm (0.015 in.) dia., Annealed, Temper: soft
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberP15I017 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043996.8V
Lot NumberN15I008 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043996.8V
Lot NumberP22D017 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043996.8V
Lot NumberI08N28 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043996.8V
Lot NumberL12X030 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number043996.8V

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum wire, 0.375mm (0.015in) dia, annealed, 99.95% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.