missing translation for 'onlineSavingsMsg'
Learn More
Molybdenum wire, 1.14mm (0.045in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals Product Code.: 15486695

Molybdenum wire, 1.14mm (0.045in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals

Product Code. 15486695
25 m
Click to view available options
Quantity:
10 m
25 m
5 m
This item is not returnable. View return policy

Product Code. 15486695

Brand: Thermo Scientific Alfa Aesar 045054.G9

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificatori chimici

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifica

Quantity 25 m
Percent Purity 99.9%
Weight ≈10.5g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Molybdenum wire, 1.14mm (0.045 in.) dia., Annealed

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT11K022 Certificate TypeCertificate of Analysis Date10/01/2025 Catalog Number045054.G5,045054.G6,045054.G9
Lot NumberT11K021 Certificate TypeCertificate of Analysis Date02/10/2024 Catalog Number045054.G5,045054.G6,045054.G9
Lot NumberL14S004 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045054.G5,045054.G6,045054.G9
Lot NumberW17F025 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045054.G5,045054.G6,045054.G9
Lot NumberB21Y033 Certificate TypeCertificate of Analysis Date12/11/2021 Catalog Number045054.G5,045054.G6,045054.G9

RUO – Research Use Only

Correzione del contenuto del prodotto

Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.

Titolo del prodotto
Molybdenum wire, 1.14mm (0.045in) dia, annealed, 99.9% (metals basis), Thermo Scientific Chemicals >

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.

Grazie per averci aiutato a migliorare il nostro sito web. Il vostro feedback è stato inviato