missing translation for 'onlineSavingsMsg'
Learn More

Molybdic Acid, Honeywell Fluka™

Product Code. 15672300
Click to view available options
Quantity:
100 g
500 g

ACS Reagent,≥85.0% MoO3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00036287
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
PubChem CID 82208
ChEBI CHEBI:25371
IUPAC Name dihydroxy(dioxo)molybdenum
SMILES O[Mo](O)(=O)=O

Specifications

Quantity 100 g
UN Number NONH for all modes of transport
Percent Purity ≥85% (MoO3 basis)
Grade ACS Reagent
Density 3.1g/mL at 25°C; Vapor Density: 6.2 (vs air)
Chemical Name or Material Molybdic Acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'