missing translation for 'onlineSavingsMsg'
Learn More
Molybdovanadic Reagent, European Pharmacopoeia Grade, Fisher Chemical™ Product Code.: 12960464
compliance-icons compliance-icons

Molybdovanadic Reagent, European Pharmacopoeia Grade, Fisher Chemical™

Product Code. 12960464
100 mL
Click to view available options
Quantity:
100 mL
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 12960464

Brand: Fisher Chemical RP 129 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Quantity 100 mL

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1080215 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 129 100
Lot Number1000233 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 129 100
Lot Number1002249 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 129 100
Lot Number1002831 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 129 100
Lot Number1001268 Certificate TypeCertificate of Analysis Date04/03/2025 Catalog NumberRP 129 100
compliance-icons compliance-icons
Product Identifier
  • Molybdovanadic Reagent. European Pharmacopoeia Ref: 1056700. (RP 129)
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Carcinogenicity Category 2
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdovanadic Reagent, European Pharmacopoeia Grade, Fisher Chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.