Learn More
Molymod™ Molecular Model Organic Teacher Set
Brand: Molymod
Code : 02
Additional Details : Weight : 0.20000kg
Specifications
Organic Set, Teacher | |
40 Hydrogen (l), 38 Carbon (I, III, IV, V), 12 Oxygen (ll), 4 Nitrogen (lV), 2 Sulphur (ll, lV), 4 Phosphorus (lV), 8 Halogen (I), 3 Metal (I, II), 18 Lone Pair, 20 Paddles (6 Pink, 6 Purple, 6 Beige), Links 55 Medium (Grey), 25 Long Flexible (gray), 25 long flexible (gray), 60 short (white), 1 short link remover tool | |
111 Atoms |