missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monomeric Cyanine Nucleic Acid Stains

Monomeric Cyanine Nucleic Acid stains are five spectrally distinct dyes for ultrasensitive fluorescence detection of nucleic acid in imaging and flow cytometry. They have bright fluorescence signals, low background, and strong binding affinity with nucleic acid.

Brand:  Invitrogen™ P3581

295.64 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 10482852

  • £389.00 / 1mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

For Research Use Only. All usage must comply with product instructions.
 
This item is not returnable. View return policy

Description

Description

Monomeric Cyanine Nucleic Acid stains allow ultrasensitive fluorescence detection of double-stranded nucleic acids. These high-affinity dyes have low background and bright fluorescence when bound to nucleic acid. They are ideally suited for nuclear staining in flow cytometry and fluorescence microscopy applications in fixed cells, along with nucleic acid detection on solid supports and in electrophoresis applications.

These five spectrally distinct monomeric cyanine nucleic acid stains are high sensitivity fluorescent probes for nucleic acid staining. The carbocyanine monomers have very strong binding affinity for dsDNA, with dissociation constants in the micromolar range. These dyes each comprise a single cyanine dye and a cationic side chain. The carbocyanine monomers are spectrally analogous to the corresponding dimeric cyanine dyes; however, with only two positive charges and one intercalating unit, these dyes exhibit somewhat reduced affinity for nucleic acids relative to the cyanine dimers.

Features of the monomeric cyanine nucleic acid stains:
High affinity—strong binding affinity for dsDNA with dissociation constants in the micromolar range
Low background—negligible fluorescence when not bound to nucleic acids
Large fluorescence enhancement—20- to 1800-fold fluorescence enhancements upon binding to DNA
Bright fluorescence—high extinction coefficients and fluorescence quantum yields resulting in very bright fluorescence signals

Like their dimeric counterparts, these monomeric cyanine dyes are typically impermeant to viable cells and can be used as dead cell indictors, although YO-PRO-1 (Cat. No. Y3603) can be permeant to apoptotic cells, providing a convenient indicator of apoptosis. The monomeric cyanine stains have wide applicability due to their low background and bright fluorescence. Uses include staining of nucleic acids on solid supports, prestaining of samples for gel or capillary electrophoresis, viability detection, and counterstaining in multiple-label experiments.

The fluorescence spectra of the five monomeric cyanine nucleic acid stains cover the entire visible wavelength range and each dye differs in extinction coefficient and quantum yield. These dyes may also be used with ultraviolet trans- or epi-illuminator excitation sources. TO-PRO-1, PO-PRO-1, and YO-PRO-1 dyes exhibit secondary ultraviolet excitation peaks near 275 nm. The monomeric cyanine nucleic acid stains exhibit large degrees of fluorescence enhancement upon binding to DNA (or RNA), varying from 400- to 1800-fold for the “1-series” and 20- to 200-fold for the “3-series.” Consequently, the fluorescence of unbound dye is negligible under most experimental detection conditions.

The monomeric cyanine nucleic acid stains are supplied in a unit size of 1 mL as 1 mM solutions in dimethylsulfoxide (DMSO).

PO-PRO™-1 Iodide (435/455)
The blue-fluorescent PO-PRO-1 stain has excitation/emission maxima ∼435/455 nm when bound to nucleic acid. It can be excited by ultraviolet (UV) excitation sources and the He-Cd 442 nm laser.

YO-PRO™-1 Iodide (491/509)
The green-fluorescent YO-PRO-1 stain has excitation/emission maxima ∼491/509 nm when bound to nucleic acid. It can be excited by ultraviolet (UV) excitation sources and the Ar 488 nm laser. YO-PRO-1 stain has been used to identify apoptotic cells. Apoptotic cells become permeant to YO-PRO-1, but remain impermeant to propidium iodide (Cat. No. P3566) and other dead cell stains. Live cells are not stained with YO-PRO-1, allowing them to be used in subsequent experiments. The YO-PRO-1 nucleic acid stain is also included as an apoptosis indicator in the Vybrant Apoptosis Assay Kit #4 (Cat. No. V13243) and Chromatin Condensation/Membrane Permeability/Dead Cell Apoptosis Kit (Cat. No. V23201).

TO-PRO™-1 Iodide (515/531)
The green-fluorescent TO-PRO-1 stain has excitation/emission maxima ∼515/531 nm when bound to nucleic acid. It can be excited by ultraviolet (UV) excitation sources and the Ar 514 nm laser.

YO-PRO™-3 Iodide (612/631)
The orange-fluorescent YO-PRO-3 stain has excitation/emission maxima ∼612/631 nm when bound to nucleic acid. It can be excited by the He-Ne 594 nm laser.

TO-PRO™-3 Iodide (642/661)
The far-red-fluorescent TO-PRO-3 stain has excitation/emission maxima ∼642/661 nm with fluorescence similar to Alexa Fluor 647 or Cy5 dyes. The long-wavelength fluorescence of TO-PRO-3 stain is well separated from that of commonly used fluorophores, such as the Alexa Fluor dyes, Oregon Green, fluorescein (FITC), rhodamine (TRITC), Texas Red, coumarin (AMCA), Marina Blue, and Pacific Blue dyes. Additionally, long-wavelength light–absorbing dyes such as TO-PRO-3 stain have the advantage that their fluorescence is usually not obscured by the autofluorescence of tissues.

TO-PRO-3 stain is useful as a nuclear counterstain and dead cell indicator in fluorescence microscopy and flow cytometry, and is among the highest-sensitivity probes for nucleic acid detection. TO-PRO-3 gives strong and selective staining of the nucleus in cultured cells and in paraffin sections. Simultaneous labeling with a green-fluorescent SYTO dye and cell-impermeant TO-PRO-3 stain is frequently used to assess cell viability. TO-PRO-3 has a much higher extinction coefficient than DNA-bound propidium iodide and has been used for unique applications such as detecting microbial cells and microorganisms in soil environments.

TO-PRO-3 is also available as room-temperature-stable, ready-to-use solutions:
• NucRed™ Dead 647 ReadyProbes™ Reagent (Cat. No. R37113)
• TO-PRO™ 3 Ready Flow™ Reagent (Cat. No. R37170)

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Blue
455 nm
Tube
Fluorescence Microscope
Fluorescence
435 nm
Room Temperature
Nucleic Acid Stain
Cell-impermeant
Store in freezer at -5°C to -30°C and protect from light.
Cell-impermeant
Fluorescent Labeling, Flow Cytometry
1 mL
PO-PRO™-1 Iodide (435/455), 1 mM Solution in DMSO
PO-PRO™
Fluorescent Dye
Nucleic Acids, Nucleus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.