missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkey Decorin ELISA Kit

Monkey Decorin quantitates monkey Decorin in serum, plasma, supernatant.

£530.00 - £2215.00

Specifications

Accession Number bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product
Assay Range 5 to 1,200 pg/mL
Assay Sensitivity 5 pg/mL
Conjugate Biotin
Product Type ELISA
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17125819
View Documents Promotion Details
Invitrogen™
EP22RBX5
5 x 96 Tests
£2215.00
Each
View alternative products
Save Up to 
»
Estimated Shipment: 04-11-2024
to see stock.
Add to basket
 
17174943
View Documents Promotion Details
Invitrogen™
EP22RB
96 Tests
£530.00
Pack of 96
View alternative products
Save Up to 
»
Estimated Shipment: 05-11-2024
to see stock.
Add to basket
 
Description

Description

Monkey Decorin quantitates monkey Decorin in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant monkey Decorin.

Decorin is a member of the small, leucine-rich proteoglycan (SLRP) family along with biglycan. Found in the interterritorial region of cartilage, decorin interacts with collagen and other matrix proteins to regulate the cartilaginous matrix. It has been shown to interact with fibronectin, thrombospondin, C1q, epidermal growth factor receptor (EGFR), and TGF-b. It is believed that decorin also plays a role in the cell cycle of chondrocytes. The decorin neoepitope fragment has a sequence of PKTLQE, and is generated by MMP-3 or ADAMTS-4 cleavage at Glu154-Leu155 of decorin core protein.
Specifications

Specifications

bone proteoglycan II; CSCD; DC; Dcn; DCN protein; DCN1C; Decorin; decorin isoform a preproprotein; Dermatan sulfate proteoglycan-II; dermatan sulfate proteoglycan-II (decorin); dermatan sulphate proteoglycans II; DSPG; DSPG2; mDcn; PG40; PGII; PGS2; PG-S2; proteoglycan core protein; proteoglycan core protein; binds type I collagen fibers and TGF-beta; decoron; similar to bone proteoglycan II; similar to decorin; SLRR1B; small leucine-rich protein 1 B; unnamed protein product
5 pg/mL
ELISA
Colorimetric Microplate Reader
<10%
HRP
Supernatant, 50 μL; Plasma, 50 μL; Serum, 50 μL
Decorin
4 hr 45 min
5 to 1,200 pg/mL
Biotin
Plasma, Serum, Supernatant
<12%
Pre-coated 96 well plate, Standard, Assay Diluent concentrate, Biotinylated Detection Antibody, SAV-HRP, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Research Use Only
2°C to 8°C
1 hr 20 min
Videos
SDS
Documents

Documents

Product Certifications