missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Monkey MRP2 Vesicles

Product Code. 10430260
Click to view available options
Quantity:
0.5 mL
Unit Size:
Each
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Product Code. 10430260

Brand: Invitrogen™ GM0018

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Monkey MRP2 Vesicles are insect derived purified plasma membranes with inserted MRP2 transport protein (multidrug resistance-associated protein, ABCC2). Our Monkey MRP2 Vesicles can be used to evaluate test compounds and drug interactions with MRP2 in in vitro assays.

TRUSTED_SUSTAINABILITY

Specifications

Physical Form Frozen
Quantity 0.5 mL
Purity Research Grade
Host Species Monkey
Content And Storage Store at -80°C.
Format Vial
Shipping Condition Dry Ice

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.