missing translation for 'onlineSavingsMsg'
Learn More

Mono-Poly™ Resolving Medium

Mono-Poly™ Resolving Medium separates mononuclear and polymorphonuclear leukocytes from whole human blood in two distinct bands.

Brand:  MP Biomedicals 091698049


Product Code. 17387469

  • £155.00 / 100mL

Please to purchase this item. Need a web account? Register with us today!

Currently Unavailable: We have no information when this product will be available
View alternative products
Explore more special offers
This item is not returnable. View return policy

Description

Description

Mono-Poly is highly versatile and can be used for a variety of research and clinical applications such as histocompatibility testing, lymphocyte studies, in vitro cell mediated immunity assays, or any procedure requiring pure white blood cell preparation

Product provides:

  • Separation of mononuclear and polymorphonuclear leukocytes in two distinct bands
  • Greater than 95% cell viability
  • Easy, single step procedure
  • No sample dilution requirement or lysis of red blood cells
TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 mL
Mono-Poly™ Resolving Medium
Product Suggestions

Product Suggestions

Videos
Safety and Handling
Show More
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Mono-Poly™ Resolving Medium > Mono-Poly Resolving Medium, 100mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.