missing translation for 'onlineSavingsMsg'
Learn More

mono-tert-Butyl malonate, 95%, Thermo Scientific Chemicals

Product Code. 11354142
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40052-13-9
Molecular Formula C7H12O4
Molecular Weight (g/mol) 160.17
InChI Key NGGGZUAEOKRHMA-UHFFFAOYSA-N
Synonym 3-tert-butoxy-3-oxopropanoic acid, malonic acid mono-tert-butyl ester, mono-tert-butyl malonate, 2-tert-butoxycarbonyl acetic acid, tert-butyl hydrogen malonate, mono-t-butyl malonate, pubchem13754, malonicacidmono-tert-butylester, malonic acid mono t-butyl ester
PubChem CID 545853
IUPAC Name 3-[(2-methylpropan-2-yl)oxy]-3-oxopropanoic acid
SMILES CC(C)(C)OC(=O)CC(=O)O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 19°C to 20°C
Color Colorless
Boiling Point 90°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Quantity 10 g
Formula Weight 160.17
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material mono-tert-Butyl malonate
Show More Show Less
compliance-icons
Product Identifier
  • mono-tert-Butyl malonate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'