missing translation for 'onlineSavingsMsg'
Learn More

Velp Scientifica™ MonoAluBlock™ Block with Seventeen Positions (28 x 24 mm)

For Use With: AREX-6 Hotplate Stirrers

Brand:  Velp Scientifica™ A00000338

Product Code. 15865211

  • £104.00 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Used to perform numerous reactions reducing time and bench space
  • For use with Velp Series 6 Hotplate Stirrers
Specifications

Specifications

28 mm
24 mm
Block
AREX-6 Hotplate Stirrers
17 Positions
43 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Velp Scientifica™ MonoAluBlock™ Block with Seventeen Positions (28 x 24 mm) > Holds: 17 x 28 mm Vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.